Nitrile gloves, Showa 6110PF
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6110PFS 6110PFL 6110PFXL 6110PFXXL 6110PFM 6110PFXS
112-5637EA 221 DKK
112-5637 112-5639 112-5640 112-5641 112-5638 BEST6110PFXS
Nitrile gloves, Showa 6110PF
Handsker
These disposable gloves are powder-free and made of 100% nitrile. By using Showa’s Eco Best Technology®, the 6110PF’s are completely eco-friendly, breaking down by 82% in just 386 days when tested in a laboratory.
  • Zero natural rubber latex proteins
  • Rolled cuff
  • Decreased risk of allergies
  • High performance of protection
  • 'Second skin' fit and feel
Eco Best Technology® (EBT) is a breakthrough innovation that makes the gloves completely, without affecting shelf life or sacrificing performance. When disposed in landfills, microorganisms consume, metabolise and break down EBT materials into three natural compounds - organic soil, methane and carbon dioxide. This creates fertiliser, leaving zero waste behind.
Typical applications include: Public utilities, janitorial, healthcare, pharmaceutical, landscaping, food processing, automotive and agriculture.
Certificeringer: EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS; Cat. III
Food contact approved. GreenCircle certified. Biodegradation test results acc. to the norm ASTM D5526.
Emballering: The dispenser is made of 85% recycled cardboard, the outer carton is made of 90% recycled material.