Contact plates
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
200208 200218
DELT200208EA 1210 DKK
DELT200208 DELT200218
Contact plates
Dyrkningsplader Petriskåle
Manufactured from transparent PS, these contact plates are for use in the pharmaceutical industry and in hospital environments for determining bacterial contamination of surfaces such as the skin, operating tables, refrigerated gondolas, and work surfaces.
  • Aseptic production
  • Moulded grid makes it possible to identify the detected contamination per cm² and it facilitates the counting of colonies
  • Shape of the lid makes stacking perfectly stable during transport and incubation, and saves space on work surfaces
Ø×H: 65,7×14,7 mm