Protein delivery reagent, Ab-DeliverIN™
Leverandør: OZ BIOSCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AI20100 AI20250 AI20500 AI21000
OZBIAI20100EA 1670 DKK
OZBIAI20100 OZBIAI20250 OZBIAI20500 OZBIAI21000
Protein delivery reagent, Ab-DeliverIN™
Transfektionsreagenser
Ab-DeliverIN™ transfection reagent is dedicated to the intracellular delivery of fully functional antibodies. It opens up new fields of investigation in proteomics, for instance to elucidate complex molecular mechanisms or to design new potential therapies.
  • Intracellular delivery of functionally active antibody
  • Highly efficient in many primary cells and cell lines
  • Serum compatible
  • Non toxic
  • Straightforward protocol and ready to use reagent
Store Ab-DeliverIN™ at 4 °C and FITC labeled IgG at –20 °C.
Leveringsinformation: Shipped at room temperature.