ESD occupational shoes, clogs, HACCP, Dynamic, 37361
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37361/45 37361/46 37361/47 37361/40 37361/41 37361/42 37361/43 37361/48 37361/44 37361/37 37361/38 37361/39 37361/35 37361/36
ABEB37361/45EA 446 DKK
ABEB37361/45 ABEB37361/46 ABEB37361/47 ABEB37361/40 ABEB37361/41 ABEB37361/42 ABEB37361/43 ABEB37361/48 ABEB37361/44 ABEB37361/37 ABEB37361/38 ABEB37361/39 ABEB37361/35 ABEB37361/36
ESD occupational shoes, clogs, HACCP, Dynamic, 37361
Sko og støvler
ESD clog made from microfibre with a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
  • Respiration-absorbent lining
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
  • Washable at 30 °C
For use in kitchens.
Available in different colours.
Certificeringer: EN 61340 (ESD); HACCP; EN ISO 20347:2012; OB, FO, E, A, SRC.