Tubing connectors for threaded connection caps
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01350000 01350001 01360001 01360000
201-1032EA 16.9 DKK
201-1032 201-1193 201-1034 201-1033
Tubing connectors for threaded connection caps
Fittings og stik
PP with silicone seals.
  • Suitable for threaded connection caps with GL 14 and GL 18 threads
  • Robust
Bestillingsoplysninger: Caps can be ordered separately.