SPE columns, acrylamide analysis, CHROMABOND® Carbon A
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
730165 730167
MANA730165EA 2040 DKK
MANA730165 MANA730167
SPE columns, acrylamide analysis, CHROMABOND® Carbon A
Kromatografikolonner
CHROMABOND® Carbon A is a special phase for SPE enrichment of acrylamide from water.
  • Activated and highly porous carbon
  • Spherical particles
  • High specific surface
Application: Enrichment of acrylamide from drinking water according to DIN EN ISO 38413-6.