TLC adsorbents, Polyamide 6
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
816610.1 816620.1
MANA816610.1EA 10530 DKK
MANA816610.1 MANA816620.1
TLC adsorbents, Polyamide 6
Kromatografi sorbenter TLC Sorbenter
Polyamide 6 adsorbents for thin layer chromatography.
  • High-quality polyamide
  • Special separation mechanism (hydrogen bonds to amide groups)
  • Especially recommended for natural compounds
  • Available with or without fluorescence indicator