COD standard solutions, Tintometer®
Leverandør: Lovibond
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2420804 2420803 2420805
LOVI2420804EA 422 DKK
LOVI2420804 LOVI2420803 LOVI2420805
COD standard solutions, Tintometer®
Standarder Miljø analyse standarder
Standard solutions are solutions with a defined concentration and are provided to check the operation methods and devices of the cuvette tests as well as the condition of optical filters and the instrument.