Spectinomycin dihydrochloride pentahydrate 95 Titrimetric, BioChemica®
Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate
A3834.0005 A3834.0010
A3834.0005EA 1800 DKK
A3834.0005 A3834.0010
Spectinomycin dihydrochloride pentahydrate 95 Titrimetric, BioChemica®
Spectinomycin dihydrochloride pentahydrate
Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Molekylvægt: 495.35 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150886
CAS-nummer: 22189-32-8
EINECS: 244-554-3
Bestil nu
Specifikationer Testresultater
Assay (Titrimetric) Min. 95%