Pans for autosamplers, Diamond™ DSC
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N5190788 N5370550 B0182902 03190025 03190029 02190042
PERCN5190788EA 8710 DKK
PERCN5190788 PERCN5370550 PERCB0182902 PERC03190025 PERC03190029 PERC02190042
Pans for autosamplers, Diamond™ DSC
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Used for any non-metal, i.e. ceramics, coal, soils, or minerals. Can also be used for any material that reacts with aluminum or which must be heated over 600° C.
Not for use with Diamond DSC,PYRIS 1 DSC, or DSC 7 Autosampler.