ESD safety boots, lace-up, Safeguard, Jonte
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
78691/40 78691/43 78691/38 78691/45 78691/44 78691/39 78691/47 78691/41 78691/42 78691/37 78691/46
ELTN78691/40EA 873 DKK
ELTN78691/40 ELTN78691/43 ELTN78691/38 ELTN78691/45 ELTN78691/44 ELTN78691/39 ELTN78691/47 ELTN78691/41 ELTN78691/42 ELTN78691/37 ELTN78691/46
ESD safety boots, lace-up, Safeguard, Jonte
Sko og støvler
Metal-free ESD winter boot made from cowhide leather/hydrophobized nubuck leather and hydrophobized textile material CORDURA®, a full, padded bellows tongue, reflective materials and a PU/PU tread sole SAFEGUARD.
  • Warm lining
  • Full-length warm fleece ESD inlay sole
  • Anatomically shaped insole supports the natural heel-to-toe movement of the foot
  • PU scuff cap protects the leather upper
  • Combination of laces and zipper
  • Protective composite toecap (200 J)
  • Metal-free puncture resistant midsole
  • Extra-wide version, Metal-free equipment
Suitable for damp and cold conditions.
Colour: Black/grey
Certificeringer: EN 61340 (ESD); EN ISO 20345:2011; S3, SRC, CI