ESD safety ankle boots, lace-up, Wellmax, Lakers XXT
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
769711/36 769711/37 769711/38 769711/39 769711/40 769711/41 769711/42 769711/43 769711/44 769711/45 769711/46 769711/47 769711/48 769711/35 769711/49
ELTN769711/36EA 716 DKK
ELTN769711/36 ELTN769711/37 ELTN769711/38 ELTN769711/39 ELTN769711/40 ELTN769711/41 ELTN769711/42 ELTN769711/43 ELTN769711/44 ELTN769711/45 ELTN769711/46 ELTN769711/47 ELTN769711/48 ELTN769711/35 ELTN769711/49
ESD safety ankle boots, lace-up, Wellmax, Lakers XXT
Sko og støvler
Metal- and leather-free ESD safety ankle boots made from microfibre upper with mesh inserts, a full padded bellows tongue plus a dual-density PU/PU sole (Wellmaxx Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Anatomically-shaped insole and ankle support for improved stability
  • Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC
Bestillingsoplysninger:
Bestil nu