ESD safety ankle boots, lace-up, Malvin Blue
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
769541/41 769541/42 769541/43 769541/44 769541/45 769541/46 769541/47 769541/40 769541/48
ELTN769541/41EA 783 DKK
ELTN769541/41 ELTN769541/42 ELTN769541/43 ELTN769541/44 ELTN769541/45 ELTN769541/46 ELTN769541/47 ELTN769541/40 ELTN769541/48
ESD safety ankle boots, lace-up, Malvin Blue
Sko og støvler
ESD ankle boots made from hydrophobised nubuck leather with mesh inserts, a full padded bellows tongue plus a dual-density TPU/PU sole (Maxximo).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Sportive ESD)
  • Anatomically-shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Abrasion resistant scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC.