ESD safety ankle boots, lace-up, Wellmax, Miles
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
769451/40 769451/41 769451/44 769451/45 769451/46 769451/42 769451/43 769451/47 769451/48
ELTN769451/40EA 850 DKK
ELTN769451/40 ELTN769451/41 ELTN769451/44 ELTN769451/45 ELTN769451/46 ELTN769451/42 ELTN769451/43 ELTN769451/47 ELTN769451/48
ESD safety ankle boots, lace-up, Wellmax, Miles
Sko og støvler
ESD safety ankle boots made from cowhide leather upper and hydrophobised nubuck leather, a full padded bellows tongue and a dual-density TPU/PU sole WELLMAXX.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • ESD soft-fleece insole with integrated omissions
  • Anatomically shaped insole and ankle support for improved stability
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC