ESD safety ankle boots, lace-up, Biomex Dynamics, Till Boa
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
176651/36 76651/37 76651/38 76651/39 76651/40 76651/41 76651/42 76651/43 76651/44 76651/45 76651/46 76651/47 76651/48 276651/49
ELTN176651/36EA 850 DKK
ELTN176651/36 ELTN76651/37 ELTN76651/38 ELTN76651/39 ELTN76651/40 ELTN76651/41 ELTN76651/42 ELTN76651/43 ELTN76651/44 ELTN76651/45 ELTN76651/46 ELTN76651/47 ELTN76651/48 ELTN276651/49
ESD safety ankle boots, lace-up, Biomex Dynamics, Till Boa
Sko og støvler
ESD safety ankle boots made from water resistant cowhide leather upper, a full padded bellows tongue an extremely flexible, ergonomic, S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
  • Full length fleece inlay sole (ESD Pro), black
  • Replaceable insole; orthopaedic insoles possible
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Lace- and buckle-free Boa® system can be opened and closed with just one hand
  • TPU scuff cap protects the upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, HI, HRO.