ESD safety ankle boots, lace-up, L10, Emotion
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761091/39 761091/40 761091/41 761091/42 761091/44 761091/37 761091/38 761091/43 761091/45 761091/46 761091/47 761091/48
114-8813EA 701 DKK
114-8813 114-8814 114-8815 114-8816 114-8818 114-8811 114-8812 114-8817 114-8819 114-8820 114-8821 114-8822
ESD safety ankle boots, lace-up, L10, Emotion
Sko og støvler
Lightweight, vintage and casual style, environmental friendly manufactured ESD safety ankle boot made from hydrophobised nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
  • Fabric lining Bioactive
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO beige
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Beige
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC