ESD safety shoes, lace-up, Nova, Mila
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
74720/35 74720/36 74720/37 74720/38 74720/39 74720/40 74720/41 74720/42
ELTN74720/35EA 574 DKK
ELTN74720/35 ELTN74720/36 ELTN74720/37 ELTN74720/38 ELTN74720/39 ELTN74720/40 ELTN74720/41 ELTN74720/42
ESD safety shoes, lace-up, Nova, Mila
Sko og støvler
Leather-free, lightweight ESD safety shoe made from microfibre/mesh material, a padded tongue plus a slip resistant, dual-density PU/PU sole (Nova).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Ladies ESD)
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in dry areas.
Colour: Grey/turquoise
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC