ESD safety shoes, lace-up, Wellmax, Reaction XXT
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729701/36 729701/37 729701/38 729701/39 729701/40 729701/41 729701/42 729701/43 729701/44 729701/45 729701/46 729701/47 729701/48
ELTN729701/36EA 708 DKK
ELTN729701/36 ELTN729701/37 ELTN729701/38 ELTN729701/39 ELTN729701/40 ELTN729701/41 ELTN729701/42 ELTN729701/43 ELTN729701/44 ELTN729701/45 ELTN729701/46 ELTN729701/47 ELTN729701/48
ESD safety shoes, lace-up, Wellmax, Reaction XXT
Sko og støvler
Metal-free ESD safety shoes made from waxy cowhide leather, full, padded bellows tongue plus a dual-density PU/PU sole (Wellmaxx Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole ESD PRO black
  • Anatomically-shaped insole and ankle support for improved stability
  • Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
  • Abrasion-proof scuff cap
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC