ESD safety shoes, lace-up, Maddox S3
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729561/35 729561/36 729561/37 729561/38 729561/39 729561/40 729561/41 729561/42 729561/43 729561/44 729561/45 729561/46 729561/47 729561/48
ELTN729561/35EA 671 DKK
ELTN729561/35 ELTN729561/36 ELTN729561/37 ELTN729561/38 ELTN729561/39 ELTN729561/40 ELTN729561/41 ELTN729561/42 ELTN729561/43 ELTN729561/44 ELTN729561/45 ELTN729561/46 ELTN729561/47 ELTN729561/48
ESD safety shoes, lace-up, Maddox S3
Sko og støvler
Leather-free ESD safety shoes made from hydrophobised microfibre and hydrophobised textile material, a full padded bellows tongue plus a dual-density TPU/PU sole (Maxximo).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (Sportive ESD)
  • Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC