ESD safety shoes, lace-up, Senex
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1728321/35 1728321/36 1728321/37 728321/38 728321/39 728321/40 728321/41 728321/42 728321/43 728321/44 728321/45 728321/46 728321/47
ELTN1728321/35EA 656 DKK
ELTN1728321/35 ELTN1728321/36 ELTN1728321/37 ELTN728321/38 ELTN728321/39 ELTN728321/40 ELTN728321/41 ELTN728321/42 ELTN728321/43 ELTN728321/44 ELTN728321/45 ELTN728321/46 ELTN728321/47
ESD safety shoes, lace-up, Senex
Sko og støvler
Metal- and leather-free, sporty and lightweight, ESD safety shoe made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC