ESD safety shoes, lace-up, Safety-Grip, Alessio Blue
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
727541/36 727541/37 727541/38 727541/39 727541/40 727541/41 727541/42 727541/43 727541/44 727541/45 727541/46 727541/47 727541/48 727541/49 727541/50
ELTN727541/36EA 492 DKK
ELTN727541/36 ELTN727541/37 ELTN727541/38 ELTN727541/39 ELTN727541/40 ELTN727541/41 ELTN727541/42 ELTN727541/43 ELTN727541/44 ELTN727541/45 ELTN727541/46 ELTN727541/47 ELTN727541/48 ELTN727541/49 ELTN727541/50
ESD safety shoes, lace-up, Safety-Grip, Alessio Blue
Sko og støvler
ESD safety shoe made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-desity PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for useMore Product Information