ESD safety shoes, lace-up, Lakers Black
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
724451/40 724451/43 724451/48 724451/49 724451/41 724451/42 724451/37 724451/38 724451/39 724451/44 724451/45 724451/46 724451/47
ELTN724451/40EA 634 DKK
ELTN724451/40 ELTN724451/43 ELTN724451/48 ELTN724451/49 ELTN724451/41 ELTN724451/42 ELTN724451/37 ELTN724451/38 ELTN724451/39 ELTN724451/44 ELTN724451/45 ELTN724451/46 ELTN724451/47
ESD safety shoes, lace-up, Lakers Black
Sko og støvler
Metal- and leather-free ESD safety shoes made from microfibre upper with mesh inserts, a full padded bellows tongue and a dual-density PU/PU sole FUN.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • Anatomically shaped insole and ankle support for improved stability
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC