ESD safety shoes, sandals, Velcro® fastening, Sid
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72131/38 72131/39 72131/40 72131/41 72131/42 72131/43 72131/44 72131/45 72131/46 72131/47
ELTN72131/38EA 611 DKK
ELTN72131/38 ELTN72131/39 ELTN72131/40 ELTN72131/41 ELTN72131/42 ELTN72131/43 ELTN72131/44 ELTN72131/45 ELTN72131/46 ELTN72131/47
ESD safety shoes, sandals, Velcro® fastening, Sid
Sko og støvler
100% metal- and leather-free, sporty and lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised Cordura® (tear and abrasion resistant, dirt and water repellent), a padded tongue and a slip resistant dual-density TPU/PU sole Trainers.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length fleece inlay sole ESD PRO black
  • TPU scuff cap protects the upper
  • Metal-free, puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moistureMore Product Information