Safety Boots, Lace-Up, Safety-Grip, Joris GTX
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
68461/48 68461/39 68461/40 68461/41 68461/42 68461/43 68461/44 68461/45 68461/46 68461/47
ELTN68461/48EA 895 DKK
ELTN68461/48 ELTN68461/39 ELTN68461/40 ELTN68461/41 ELTN68461/42 ELTN68461/43 ELTN68461/44 ELTN68461/45 ELTN68461/46 ELTN68461/47
Safety Boots, Lace-Up, Safety-Grip, Joris GTX
Sko og støvler
Durable ESD safety shoes made from robust, breathable cowhide leather and hydrophobised nubuck leather, a full padded bellows tongue plus a temperature resistant, dual-density rubber/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Gore-Tex® climate membrane
  • Full length inlay sole (Basic), grey
  • Antistatic soft fleece insole
  • Anatomically-shaped insole and ankle support for improved stability
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in cold areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey/blue
Certificeringer: DGUV 112-191; EN ISO 20345; S3, SRC, CI, HI, HRO.