Safety shoes, lace-up, Safety-Grip, Renzo Winter
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
65741/36 65741/37 65741/38 65741/39 65741/40 65741/41 65741/42 65741/43 65741/44 65741/45 65741/46 65741/47 65741/48 65741/49 65741/50
ELTN65741/36EA 664 DKK
ELTN65741/36 ELTN65741/37 ELTN65741/38 ELTN65741/39 ELTN65741/40 ELTN65741/41 ELTN65741/42 ELTN65741/43 ELTN65741/44 ELTN65741/45 ELTN65741/46 ELTN65741/47 ELTN65741/48 ELTN65741/49 ELTN65741/50
Safety shoes, lace-up, Safety-Grip, Renzo Winter
Sko og støvler
Durable ESD safety shoes made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Warm woven lamb skin virgin wool lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length warm fleece inlay sole
  • Anatomically-shaped insole and ankle support for improved stability
  • PU scuff cap protects the leather upper
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
  • Extra wide version (XW)
Ideal for use in cold areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, CI