ESD safety shoes, lace-up, Wellmaxx, Logan Blue
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729425/40 729425/41 729425/42 729425/43 729425/44 729425/45 729425/46 729425/47 729425/48
ELTN729425/40EA 776 DKK
ELTN729425/40 ELTN729425/41 ELTN729425/42 ELTN729425/43 ELTN729425/44 ELTN729425/45 ELTN729425/46 ELTN729425/47 ELTN729425/48
ESD safety shoes, lace-up, Wellmaxx, Logan Blue
Sko og støvler
Lightweight and comfortable ESD safety shoes made from mesh/textile, full padded bellows tongue and collar and a dual-density TPU/PU sole WELLMAXX.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Protective composite toecap (200 J)
Ideal for use in dry areas.
Colour: Black/blue
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC