VWR® Bottles and Tube Racks, Wire, Coated
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212-0698EA 247 DKK
212-0698 212-0699 212-0700 212-0701
VWR® Bottles and Tube Racks, Wire, Coated
Stativer Flaske Stativer
Wire racks, epoxy coated.
  • Suitable for Bijou bottles and universal bottles/tubes up to 25 mm Ø
  • Mesh bottom support
  • Chemically-resistant, HDPE coated racks are good to −40 °C
Durable racks for transport or sample handling.