Earmuffs/Headset, PELTOR™ ProTac™ III
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MT13H221A MT13H221P3E MT13H220A MT13H220P3E
MMMAMT13H221AEA 1840 DKK
MMMAMT13H221A MMMAMT13H221P3E MMMAMT13H220A MMMAMT13H220P3E
Earmuffs/Headset, PELTOR™ ProTac™ III
Høreværn Støjblokerende høreværn
ProTac™ III hearing protector helps protect against harmful noise, while allowing ambient sounds to be heard at a level below 82 dB. It increases the ability to communicate with nearby colleagues, as well as hearing warning signals, approaching vehicles, or sounds in machines and processes.
  • Level-dependent function for ambient listening
  • Voice guided menu system
  • Auto power off
  • Low-battery warning at low battery level
  • To reduce the corrosion caused by sweat the electronics are located in the outer part of the cup
Available as both headband and helmet attached versions, with two different cup sizes: one with higher attenuation and one slim version with lower attenuation.
Used for work in industrial areas, construction sites, rail work and other areas.
Certificeringer: EN 352-1:2002/EN 352-3:2002, EN352-4:2001/A1:2005, EN 352-6:2002.
Leveringsinformation: Supplied with 1×headset, 2×AA batteries and 1×user instruction.