Protective hand gels, Physioderm® Proglove
Leverandør: FAWECO HAUTSCHUTZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13699-001 13699-004
FAWE13699-001EA 7330 DKK
FAWE13699-001 FAWE13699-004
Protective hand gels, Physioderm® Proglove
Hudpleje Cremer Hand Creams
Colourless, alcoholic gel protects skin from softening when wearing gloves or rubber boots.
  • Hamamelis and chitin protect the upper skin layer
  • Bisabolol and glycerine are soothing and nourishing
  • Free from perfume, preservatives, silicone and greases
Certificeringer: HACCP-compliant
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904