Autosampler vials, inserts and kits, RSA™
Leverandør: MICROSOLV
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9509S-0CV-RS 9509S-1WCP-RS 9509C-1EWCP-RS 9509C-1WCP-RS 9509C-1XWCP-RS 9509C-3EWCP-RS 9509C-3WAP-RS 9509C-3WCP-RS 9509C-3XWAP-RS 9509C-3XWCP-RS 9509C-WAV-RS 9509C-WCV-RS 9509S-1CP-RS 9509S-1EWAP-RS 9509S-1EWCP-RS 9509S-1WAP-RS 9509S-1WCP-RSA 9509S-1XWAP-RS 9509S-1XWCP-RS 9509S-3CP-RS 9509S-3EWAP-RS 9509S-3EWCP-RS 9509S-3WAP-RS 9509S-3WCPEARS 9509S-3WCP-RS 9509S-3XWAP-RS 9509S-3XWCP-RS 9509S-9WAP-RS 9509S-9WCP-RS 9509S-WAV-RS 9509S-WCV-RS 9512C-0CV-RS 9512C-0CV-T-RS 9512C-1CP-RS 9512C-1MP-RS 9512C-3CP-RS 9512C-3MP-RS 9512S-0CV-RS 9512S-1AP-RS 9512S-1CP-RS 9512S-1MP-RS 9512S-3AP-RS 9512S-3CP-RS 9512S-3MP-RS 9512S-9AP-RS 9512S-9CP-RS 9532C-0CV-RS 9532C-1CP-RS 9532C-3CP-RS 9532S-0CV-RS 9532S-1CP-RS 9532S-1CPWT-RS 9532S-3CP-RS 9532S-3CPWT-RS 9532S-WCV-WTRS 9512S-0CV-T-RS
MICS9509S-0CV-RSEA 257 DKK
MICS9509S-0CV-RS MICS9509S-1WCP-RS MICS9509C-1EWCP-RS MICS9509C-1WCP-RS MICS9509C-1XWCP-RS MICS9509C-3EWCP-RS MICS9509C-3WAP-RS MICS9509C-3WCP-RS MICS9509C-3XWAP-RS MICS9509C-3XWCP-RS MICS9509C-WAV-RS MICS9509C-WCV-RS MICS9509S-1CP-RS MICS9509S-1EWAP-RS MICS9509S-1EWCP-RS MICS9509S-1WAP-RS MICS9509S-1WCP-RSA MICS9509S-1XWAP-RS MICS9509S-1XWCP-RS MICS9509S-3CP-RS MICS9509S-3EWAP-RS MICS9509S-3EWCP-RS MICS9509S-3WAP-RS MICS9509S-3WCPEARS MICS9509S-3WCP-RS MICS9509S-3XWAP-RS MICS9509S-3XWCP-RS MICS9509S-9WAP-RS MICS9509S-9WCP-RS MICS9509S-WAV-RS MICS9509S-WCV-RS MICS9512C-0CV-RS MICS9512C-0CV-T-RS MICS9512C-1CP-RS MICS9512C-1MP-RS MICS9512C-3CP-RS MICS9512C-3MP-RS MICS9512S-0CV-RS MICS9512S-1AP-RS MICS9512S-1CP-RS MICS9512S-1MP-RS MICS9512S-3AP-RS MICS9512S-3CP-RS MICS9512S-3MP-RS MICS9512S-9AP-RS MICS9512S-9CP-RS MICS9532C-0CV-RS MICS9532C-1CP-RS MICS9532C-3CP-RS MICS9532S-0CV-RS MICS9532S-1CP-RS MICS9532S-1CPWT-RS MICS9532S-3CP-RS MICS9532S-3CPWT-RS MICS9532S-WCV-WTRS MICS9512S-0CV-T-RS
Autosampler vials, inserts and kits, RSA™
Hætteglas
RSA™ (Reduced Surface Activity) high quality and extremely consistent autosampler vials and caps due to reduced surface activity glass manufacturing and control processes. Excellent for low abundance or low dosage form analytes or samples that are pH sensitive, or basic in nature.
  • Low adsorption glass, useful for low abundance basic compounds
  • No pH change from the vial when using a high amount of water in samples
  • Virtually no surface metals prevents adducts in LCMS
  • Completely manufactured robotically, not touched by human hands
  • Reliability and conistency ensured through extreme QC testing
Available in standard 2 ml sizes, clear and amber formats, and with limited volume features when sample is limited. Improved results with LCMS, GCMS or UHPLC. All lots are tested for fit, form, cleanliness, adsorption, pH control, and metal content.
Leveringsinformation: Supplied with certificate if requested. Available in "easy purchase packs".