Anti-VIM Mouse Monoclonal Antibody [clone: V9]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM52311-R10 ILM52311-R25
VWRKILM52311-R10EA 0 DKK
VWRKILM52311-R10 VWRKILM52311-R25
Anti-VIM Mouse Monoclonal Antibody [clone: V9]
Antistoffer
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: V9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Specifikationer

Antigen symbol VIM
Antigen name Vimentin
Environmentally Preferable
Antistof type Primary
Clonality Monoclonal
Konjugation Unconjugated
Clone V9
Reactivity Human
Vært Mouse
Gen ID 7431
Isotype IgG1
ImmunoChemistry Yes
Form Ready to use
Opbevaringstemperatur 2°C - 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR