Anti-GZMB Mouse Monoclonal Antibody [clone: GRB-7]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM7026-C01 ILM7026-C05 ILM7026-C1
ILM7026-C01EA 1250 DKK
ILM7026-C01 ILM7026-C05 ILM7026-C1
Anti-GZMB Mouse Monoclonal Antibody [clone: GRB-7]
Antistoffer
Type: Primary
Antigen: GZMB
Clonality: Monoclonal
Clone: GRB-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR