Anti-Cytokeratin Mouse Monoclonal Antibody [clone: MNF116]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM2225-C01 ILM2225-C05 ILM2225-C1
ILM2225-C01EA 1160 DKK
ILM2225-C01 ILM2225-C05 ILM2225-C1
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: MNF116]
Antistoffer
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: MNF116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR