Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ILM54411-C01 ILM54411-C05 ILM54411-C1
ILM54411-C01EA 536 DKK
ILM54411-C01 ILM54411-C05 ILM54411-C1
Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Antistoffer
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: OV-TL 12/30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human