Anti-CD79A Mouse Monoclonal Antibody [clone: JCB117]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM9739-C01 ILM9739-C05 ILM9739-C1
ILM9739-C01EA 1670 DKK
ILM9739-C01 ILM9739-C05 ILM9739-C1
Anti-CD79A Mouse Monoclonal Antibody [clone: JCB117]
Antistoffer
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: JCB117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR