Anti-PTPRC Mouse Monoclonal Antibody [clone: MAC323]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM4466-C01 ILM4466-C05 ILM4466-C1
VWRKILM4466-C01EA 0 DKK
VWRKILM4466-C01 VWRKILM4466-C05 VWRKILM4466-C1
Anti-PTPRC Mouse Monoclonal Antibody [clone: MAC323]
Antistoffer
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: MAC323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Porcine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR