Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC-70]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM1313-C01 ILM1313-C05 ILM1313-C1
ILM1313-C01EA 588 DKK
ILM1313-C01 ILM1313-C05 ILM1313-C1
Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC-70]
Antistoffer
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: JC-70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Cynomolgus Monkey, Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR