Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM8403-R10 ILM8403-R25
VWRKILM8403-R10EA 0 DKK
VWRKILM8403-R10 VWRKILM8403-R25
Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]
Antistoffer
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR