Anti-CA19-9 Mouse Monoclonal Antibody [clone: 121SLE]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM1913-C01 ILM1913-C05 ILM1913-C1
ILM1913-C01EA 836 DKK
ILM1913-C01 ILM1913-C05 ILM1913-C1
Anti-CA19-9 Mouse Monoclonal Antibody [clone: 121SLE]
Antistoffer
Type: Primary
Antigen: CA19-9
Clonality: Monoclonal
Clone: 121SLE
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR