Reagents for the determination of COD for photometers
Leverandør: Palintest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL456 PL454 PL450 PL460 PL462 PL464 PL452
PALIPL456EA 1120 DKK
PALIPL456 PALIPL454 PALIPL450 PALIPL460 PALIPL462 PALIPL464 PALIPL452
Reagents for the determination of COD for photometers
Miljø test kit
Pre-dispensed liquid reagents for minimal handing with barcoded reagent labelling for ease of use in complete wastewater analysis.
  • Greater sensitivity in the lower part of the measurement range
  • Can be used with multiparameter photometer and heater block
Leveringsinformation: Supplied in 16 mm tubes.