Electronic crimping tools
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
735511 735520 735611 735620 735513 735532 735533 735613
MANA735511EA 14690 DKK
MANA735511 MANA735520 MANA735611 MANA735620 MANA735513 MANA735532 MANA735533 MANA735613
Electronic crimping tools
Tænger til Vials
These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).
  • Mobile tools for consistent and reproducible crimping results
  • Crimping pressure adjustable by the control unit on top of the tool
  • Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)
Certificeringer: CE certificate of conformity along with one year warranty.