Screw caps for screw neck vials, N 24
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
702058 702059 702061 702102 702060 702168 702130 702073 702171
MANA702058EA 946 DKK
MANA702058 MANA702059 MANA702061 MANA702102 MANA702060 MANA702168 MANA702130 MANA702073 MANA702171
Screw caps for screw neck vials, N 24
Låg Låg til kromatografirør
Bonded and ready assembled screw closures are available for these crimp neck vials.
  • Bonded closures feature a firm fit of the septum
  • Screw closure with center hole and silicone/PTFE septum are recommended for environmental analysis
  • Universal screw closure with removable protection lid for sample storage and analysis
  • Plain screw caps N 24 and replacement septa N 22 are also available