NMR sample tube caps, SP Wilmad-LabGlass
Leverandør: SP Industries
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WG-1264-5 WG-1264-10 WG-1264-3 WG-1264-4
WILMWG-1264-5EA 620 DKK
WILMWG-1264-5 WILMWG-1264-10 WILMWG-1264-3 WILMWG-1264-4
NMR sample tube caps, SP Wilmad-LabGlass
Låg Reagensglaslåg
Made of PTFE, white. NMR tube caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable caps for better spinning stability.
These caps are recommended in experiments at high-ultra high field and experiments using chloroform as reference solution. Fit precision NMR tubes only due to the tight tolerance.