VWR®, Levelling Platform with Bubble Level
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
700-0846EA 2010 DKK
700-0846 700-0847
VWR®, Levelling Platform with Bubble Level
Retortstativer
The use of levelling platforms in electrophoresis can be a big help in maintaining a uniform gel thickness and therefore a uniform and quality sample separation.
  • Uniform gel thickness
  • Consistent buffer level over the gel
  • Assures high quality separation results
The levelling platform offers 4 independent screws and an easy to read bubble level to produce a level and stable platform. With a usable surface of 36×46 cm, this platform is suitable for use with both the Mini and Midi Gel systems of the PerfectBlue™ range.