Thiosemicarbazid ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 3-Thiosemicarbazide
A14630.22 A14630.0E A14630.36
A14630.22EA 477 DKK
A14630.22 A14630.0E A14630.36
Thiosemicarbazid ≥98%
Thiosemicarbazid
Purity: 99% CAS No.: 79-19-6 Molecular Formula: CH5N3S Formula Weight: 91.14 Odour: Odourless Melting Point: ca 180° dec. Beilstein Reference: 506320 Merck Reference: 14,9361 UN No.: 2811 MDL No.: MFCD00007620
Formel: CH₅N₃S
Molekylvægt: 91.14 g/mol
Smeltepunkt: ∼ 180 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007620
CAS-nummer: 79-19-6
EINECS: 201-184-7
UN nummer: 2811
ADR: 6.1 II
Merck Index: 12,09499