3-Methoxythiophen 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
381680010
381680050
381680250
ACRO381680010EA
261
DKK
ACRO381680010
ACRO381680050
ACRO381680250
3-Methoxythiophen 98%
3-Methoxythiophene
Formel:
C₅H₆OS Molekylvægt: 114.17 g/mol Kogepunkt: 80…82 °C (65 mmHg) Smeltepunkt: –7 °C Massefylde: 1.143 g/cm³ Flammepunkt: 49 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00043894 CAS-nummer: 17573-92-1 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Clear yellow to green to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5310 to 1.5330 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...