Phenicarbazid ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B22331.14 B22331.22
B22331.14EA 913 DKK
B22331.14 B22331.22
Phenicarbazid ≥99%
Phenicarbazid

Purity: 99% CAS No.: 103-03-7 Molecular Formula: C6H5NHNHCONH2 Formula Weight: 151.17 Odour: Odourless Melting Point: 171-173° MDL No.: MFCD00007942

Formel: C₇H₉N₃O
Kogepunkt: 357 °C (1013 hPa)
Smeltepunkt: 171…173 °C
Massefylde: >1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007942
CAS-nummer: 103-03-7
EINECS: 203-072-3
Merck Index: 12,07378

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR