1-Methylpyrrol 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-Methylpyrrole, 1-Methyl-1H-pyrrole

127601000 127605000
127601000.EA 571 DKK
127601000. 127605000.
1-Methylpyrrol 99+%
1-Methylpyrrole
Formel: C₅H₇N
Molekylvægt: 81.12 g/mol
Kogepunkt: 112
Smeltepunkt: –57 °C
Massefylde: 0.91 g/cm³
Flammepunkt: 16 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005345
CAS-nummer: 96-54-8
EINECS: 202-513-7
UN nummer: 1992

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4890 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR