Pyrrol 99%, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1H-Pyrrole

157710250 157711000 157715000 157710025
ACRO157710250EA 320 DKK
ACRO157710250 ACRO157711000 ACRO157715000 ACRO157710025
Pyrrol 99%, extra pure
Pyrrole
Formel: C₄H₅N
Kogepunkt: 129…131 °C (760 mmHg)
Smeltepunkt: –23 °C
Massefylde: 0,96 g/cm³
Flammepunkt: 36 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005216
CAS-nummer: 109-97-7
EINECS: 203-724-7
UN nummer: 1992
ADR: 3,III
Merck Index: 12,08198

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20°C, 589 nm)
Water ≤0.15 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR