2-(2-Pyridyl)pyridin ≥98% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
30569.09
30569.18
30569.30
30569.09EA
454
DKK
30569.09
30569.18
30569.30
2-(2-Pyridyl)pyridin ≥98% ACS
2,2'-Bipyridyl
Purity: 98%
CAS No.: 366-18-7
Molecular Formula: C10H8N2
Formula Weight: 156.19
Flash Point: 121°(249°F)
Physical Form: Crystalline
Melting Point: 70°
Boiling Point: 272-273°
Beilstein Reference: 113089
Merck Reference: 14,3347
UN No.: 2811
MDL No.: MFCD00006212
Formel:
C₁₀H₈N₂ Kogepunkt: 272…273 °C Smeltepunkt: 70 °C Massefylde: 1,326 g/cm³ (–150 °C) Flammepunkt: 121 °C (249 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006212 CAS-nummer: 366-18-7 EINECS: 206-674-4 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,3347 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...